missing translation for 'onlineSavingsMsg'
Learn More

Caffeic Acid, (Yellowish-brown crystals),MP Biomedicals

Product Code. 11447631 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11447631 5 g 5g
11437631 1 g 1g
11457631 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11447631 Supplier MP Biomedicals Supplier No. 0210479705
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 331-39-5
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
InChI Key QAIPRVGONGVQAS-DUXPYHPUSA-N
Synonym caffeic acid, 3,4-dihydroxycinnamic acid, 3-3,4-dihydroxyphenyl acrylic acid, trans-caffeate, 3,4-dihydroxy-trans-cinnamate, 2e-3-3,4-dihydroxyphenyl prop-2-enoic acid, 3,4-dihydroxybenzeneacrylic acid, e-3-3,4-dihydroxyphenyl acrylic acid, trans-caffeic acid, 3-3,4-dihydroxyphenyl propenoic acid
PubChem CID 689043
ChEBI CHEBI:16433
IUPAC Name (E)-3-(3,4-dihydroxyphenyl)prop-2-enoic acid
SMILES C1=CC(=C(C=C1C=CC(=O)O)O)O

Specifications

Color Brown-Yellow
Quantity 5 g
Formula Weight 180.2
Physical Form Crystals
Chemical Name or Material Caffeic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.