missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calbindin D-28K Antibody (FGF2/7365), Novus Biologicals™
Mouse Monoclonal Antibody
232.00 € - 462.00 €
Specifications
| Antigen | Calbindin D-28K |
|---|---|
| Clone | FGF2/7365 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18637504
|
Novus Biologicals
NBP3-23617-100ug |
100 μg |
462.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18615954
|
Novus Biologicals
NBP3-23617-20ug |
20 μg |
232.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calbindin D-28K Monoclonal antibody specifically detects Calbindin D-28K in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Calbindin D-28K | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission, Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| 793 | |
| IgG2 κ | |
| Protein A or G purified |
| FGF2/7365 | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CAB27, CALB, calbindin, calbindin 1, (28kD), calbindin 1, 28kDa, Calbindin D28, D-28K, RTVL-H protein, Vitamin D-dependent calcium-binding protein, avian-type | |
| Recombinant fragment (around aa 7-96) of human Calbindin D-28K protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title