missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CALCB Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
196.00 € - 463.00 €
Specifications
| Antigen | CALCB |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18604132
|
Novus Biologicals
NBP2-92827-0.02ml |
0.02 mL |
196.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18692652
|
Novus Biologicals
NBP2-92827-0.1ml |
0.1 mL |
463.00 €
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CALCB Polyclonal antibody specifically detects CALCB in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| CALCB | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cardiovascular Biology, Neuroscience | |
| PBS with 50% glycerol, pH7.3. | |
| 797 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| calcitonin-related polypeptide beta, FLJ30166 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-127 of human CALCB (NP_000719.1). MGFRKFSPFLALSILVLYQAGSLQAAPFRSALESSPDPATLSKEDARLLLAALVQDYVQMKASELKQEQETQGSSSAAQKRACNTATCVTHRLAGLLSRSGGMVKSNFVPTNVGSKAFGRRRRDLQA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title