missing translation for 'onlineSavingsMsg'
Learn More

Calcitonin Polyclonal Antibody, Invitrogen™

Product Code. 11345783
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11345783 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11345783 Supplier Invitrogen Supplier No. PA516464

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA5-16464 targets Calcitonin in IHC (P) applications and shows reactivity with mouse, Non-human primate, Rat, Canine, Equine, Human, Ovine, and Porcine samples. Staining of formalin-fixed tissues requires boiling in 10mM citrate buffer pH 6.0 for 10-20 minutes followed by cooling at RT for 20 minutes. The PA5-16464 immunogen is a synthetic peptide from human calcitonin.

Calcitonin is a 32 amino acid peptide which can be demonstrated in C cells of the normal and hyperplastic thyroid. Staining for calcitonin may be used for the identification of a spectrum of C cell proliferative abnormalities ranging from C cell hyperplasia to invasive tumors. Staining for calcitonin in medullary carcinoma of the thyroid produces a fine granular pattern in the cytoplasm. Amyloid deposits within the tumor may also exhibit varying degrees of calcitonin activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calcitonin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene CALCA
Gene Accession No. P01256, P01257, P01258, P01259, P01261, P06881, P41547, P70160, Q9N0T2, Q9N0V5
Gene Alias Alpha CGRP; Alpha type CGRP; Alpha-type CGRP; CA; Cal1; CAL6; CALC; CALC1; Calca; Calcitonin; calcitonin 1; Calcitonin carboxyl-terminal peptide; calcitonin gene related peptide 1; calcitonin gene related peptide I; calcitonin gene related peptide I precursor; Calcitonin gene-related peptide; calcitonin gene-related peptide 1; Calcitonin gene-related peptide I; calcitonin precursor; calcitonin related polypeptide alpha; calcitonin/calcitonin-related polypeptide, alpha; calcitonin; calcitonin gene-related peptide 1; calcitonin-like; calcitonin-related polypeptide alpha; CCALCI; CCP; Cgrp; CGRP1; CGRP-1; CGRPI; CGRP-I; CT; Ctn; katacalcin; katacalcin (KC); KC; PCT; PDN-21; prepro-alpha-calcitonin gene-related peptide; procalcitonin; RATCAL6
Gene Symbols CALCA
Host Species Rabbit
Immunogen A synthetic peptide from human calcitonin
Purification Method Protein A
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100033906, 100294596, 100579174, 12310, 24241, 403946, 796
Target Species Canine, Equine, Human, Mouse, Non-human Primate, Ovine, Pig, Rat
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.