missing translation for 'onlineSavingsMsg'
Learn More
Learn More
calcium homeostasis modulator 2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
204.00 € - 463.00 €
Specifications
| Antigen | calcium homeostasis modulator 2 |
|---|---|
| Dilution | Western Blot 1:100 - 1:500 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18652721
|
Novus Biologicals
NBP2-92100-0.02ml |
0.02 mL |
204.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18638401
|
Novus Biologicals
NBP2-92100-0.1ml |
0.1 mL |
463.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
calcium homeostasis modulator 2 Polyclonal antibody specifically detects calcium homeostasis modulator 2 in Human, Rat samples. It is validated for Western BlotSpecifications
| calcium homeostasis modulator 2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS with 50% glycerol, pH7.3. | |
| 51063 | |
| IgG | |
| Affinity purified |
| Western Blot 1:100 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| calcium homeostasis modulator 2, calcium homeostasis modulator protein 2, FAM26Bfamily with sequence similarity 26, member B, Protein FAM26B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-323 of human CALHM2 (NP_057000.2). MAALIAENFRFLSLFFKSKDVMIFNGLVALGTVGSQELFSVVAFHCPCSPARNYLYGLAAIGVPALVLFIIGIILNNHTWNLVAECQHRRTKNCSAAPTFLLLSSILGRAAVAPVTWSVISLLRGEAYVCALSEFVDPSSLTAREEHFPSAHATEILARFPCKENPDNLSDFREEVSRRLRYESQLFGWLLIGVVAILVFLTKCLKHYCSPLSYRQEAYWAQYRANEDQLFQRTAEVHSRVLAANNVRRFFGFVALNKDDEELIANFPVEGTQPRPQWNAITGVYLYRENQGLPLYSRLHKWAQGLAGNGAAPDNVEMALLPS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title