missing translation for 'onlineSavingsMsg'
Learn More

Calcium hydride, 98% (metals basis excluding Mg), Mg <1%

Product Code. 11328788
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328788 50 g 50g
11318788 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328788 Supplier Thermo Scientific Alfa Aesar Supplier No. 041645.18
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 7789-78-8 | CaH2

Solvent in Sol-Gel precursor applications. Calcium hydride is used as an efficient drying agent for aprotic base-stable solvents like ethers and tertiary amines. It is a useful dehydrating agent in a synthesis of aldehyde enamines in high yield and purity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-78-8
Molecular Formula CaH2
Molecular Weight (g/mol) 42.09
MDL Number MFCD00010897
InChI Key FAQLAUHZSGTTLN-UHFFFAOYSA-N
IUPAC Name calcium dihydride
SMILES [CaH2]

Specifications

Melting Point 600°C (decomposition)
Color Gray
Physical Form Powder
Quantity 50 g
Assay Percent Range 98%
Solubility Information Soluble in water and alcohol. Insoluble in benzene.
Formula Weight 42.1
Sensitivity Moisture sensitive
Appearance Light gray
Density 1.9 g/cm3 at 20°C
Chemical Name or Material Calcium hydride, Mg <1%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Calcium hydride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Substance which, in contact with water, emits flammable gases Category 1
Hazard Statement
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Calcium hydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.