accessibility menu, dialog, popup

UserName

CaHH

CAS RN 7789-78-8

IUPAC Name: calcium
Synonyms: calcium calcium(0) Can calcio calcium atom kalzium
Molecular Weight (g/mol): 40.08
Molecular Formula: Ca
InChi Key: OYPRJOBELJOOCE-UHFFFAOYSA-N
SMILES: [Ca]
Quantity 
  • (6)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (5)
Physical Form 
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (7)
  • (1)
Grade 
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (7)
  • (1)
  • (7)