missing translation for 'onlineSavingsMsg'
Learn More

Calcium hydride, coarse powder, ca 92%

Product Code. 11441980
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441980 50 g 50g
11451980 250 g 250g
11431980 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11441980 Supplier Thermo Scientific Alfa Aesar Supplier No. A16242.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7789-78-8 | CaH2

Calcium hydride is used as an efficient drying agent for aprotic base-stable solvents like ethers and tertiary amines. It is a useful dehydrating agent in a synthesis of aldehyde enamines in high yield and purity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calcium chloride is used as an efficient drying agent for aprotic base-stable solvents like ethers and tertiary amines. It is a useful dehydrating agent in a synthesis of aldehyde enamines in high yield and purity.

Solubility
Soluble in water and alcohol. Insoluble in benzene.

Notes
Moisture sensitive. Reacts with water vigorously and liberates hydrogen gas. Incompatible with strong oxidizing agents, strong acids, halogens, water and alcohols.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-78-8
Molecular Formula CaH2
Molecular Weight (g/mol) 42.09
MDL Number MFCD00010897
InChI Key FAQLAUHZSGTTLN-UHFFFAOYSA-N
IUPAC Name calcium dihydride
SMILES [CaH2]

Specifications

Melting Point 600°C (decomposition)
Physical Form Coarse Powder
Quantity 50 g
UN Number UN1404
Merck Index 14,1672
Solubility Information Soluble in water and alcohol. Insoluble in benzene.
Formula Weight 42.1
Percent Purity ∼92%
Odor Odorless
Density 1.9
Chemical Name or Material Calcium hydride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.