missing translation for 'onlineSavingsMsg'
Learn More

Calcium Oil Standard in Base Oil, 1000μg/g, SPEX CertiPrep™

Product Code. 16361196 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16361196 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16361196 Supplier SPEX Certiprep Supplier No. ORGCA82Z
 Restricted Item
In Stock
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Indeholder

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ca1000μg/g

Certificeringer

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Overholdelse

GHS/OSHA Complaint

Tekniske data

Packaging Triple-leached LDPE bottle
CAS 7440-70-2, 8042-47-5
Certificate of Analysis With Certificate of Analysis.
CAS Min % 0.5, 99.5
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 0.5, 99.5
Molecular Formula Ca
MDL Number MFCD00010897 MFCD00085314
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
InChI Key OYPRJOBELJOOCE-UHFFFAOYSA-N
Type Certified Reference Material
SMILES [Ca]
IUPAC Name calcium
Solvent or Matrix Base Oil 75
Industry Type Petrochemical Testing
Molecular Weight (g/mol) 40.08
PubChem CID 5460341
Quantity 50 g
ChEBI CHEBI:29320
Grade Plasma
Concentration or Composition (by Analyte or Components) Matrix Concentration Cat. No.
Base Oil 20 1000μg/g ORG-CA8-2Z
Base Oil 75 5000μg/g ORG-CA8-4Z
Chemical Name or Material Calcium Oil Standard
Vis mere Vis mindre

Product intended for laboratory use only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.