missing translation for 'onlineSavingsMsg'
Learn More

Calcium Phenylpyruvate, TRC

Product Code. 30180344
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180344 500 mg 500mg
30179733 100 mg 100mg
30179841 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180344 Supplier Toronto Research Chemicals Supplier No. TRCC145763500MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51828-93-4
Molecular Formula (C9H7O3)2Ca
Molecular Weight (g/mol) 366.38
Synonym Benzenepropanoic acid, α-oxo-, calcium salt (2:1), Benzenepropanoic acid, α-oxo-, calcium salt (9CI), Phenylpyroracemic acid calcium salt, keto-Phenylalanine calcium, α-keto-Phenylalanine calcium, Calcium bis(3-phenylpyruvate), Calcium phenylpyruvate
IUPAC Name calcium;2-oxo-3-phenylpropanoate
SMILES [Ca+2].[O-]C(=O)C(=O)Cc1ccccc1.[O-]C(=O)C(=O)Cc2ccccc2

Specifications

Quantity 500 mg
InChI Formula InChI=1S/2C9H8O3.Ca/c2*10-8(9(11)12)6-7-4-2-1-3-5-7;/h2*1-5H,6H2,(H,11,12);/q;;+2/p-2
Formula Weight 366.0416
Percent Purity >95%
Chemical Name or Material Calcium Phenylpyruvate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.