missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caldesmon, HMW Rabbit, Clone: CALD1/1424R, enQuireBio™
Rabbit Monoclonal Antibody
Brand: EnquireBio 800-RBM4-P0
Additional Details : Weight : 0.01000kg
Specifications
Caldesmon, HMW | |
Cytoplasmic | |
CALD1/1424R | |
Unconjugated | |
NP_004333 | |
Rabbit | |
150kDa | |
Protein A/G Purified | |
RUO | |
800 | |
IgG κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
CALD1 | |
Recombinant human full-length CALD1 protein | |
Uterus, Blood vessels in all tissues, smooth muscle or leiomyosarcoma | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |