missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caldesmon, HMW Rabbit, Clone: CALD1/1424R, enQuireBio™
Rabbit Monoclonal Antibody
199.00 € - 485.00 €
Specifications
Antigen | Caldesmon, HMW |
---|---|
Clone | CALD1/1424R |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15961118
|
EnquireBio
800-RBM4-P1 |
100 μg |
485.00 €
100µg |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15951118
|
EnquireBio
800-RBM4-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Caldesmon, HMW | |
200μg/mL | |
Monoclonal | |
Rabbit | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
800 | |
Recombinant human full-length CALD1 protein | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
150kDa |
CALD1/1424R | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_004333 | |
CALD1 | |
IgG κ | |
Protein A/G Purified | |
Cytoplasmic | |
Uterus, Blood vessels in all tissues, smooth muscle or leiomyosarcoma |