missing translation for 'onlineSavingsMsg'
Learn More

Caldesmon, HMW Rabbit, Clone: CALD1/1424R, enQuireBio™

Rabbit Monoclonal Antibody

Brand:  EnquireBio 800-RBM4-P1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15961118

  • 485.00 € / 100µg
Estimated Shipment: 12-09-2024
to see stock.

Description

Description

Specifications

Specifications

Caldesmon, HMW
Cytoplasmic
CALD1/1424R
Unconjugated
NP_004333
Rabbit
150kDa
Protein A/G Purified
RUO
800
IgG κ
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with or without 0.05% BSA and 0.05% azide.
CALD1
Recombinant human full-length CALD1 protein
Uterus, Blood vessels in all tissues, smooth muscle or leiomyosarcoma
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers