missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CALHM1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00 € - 508.00 €
Specifications
| Antigen | CALHM1 |
|---|---|
| Dilution | Western Blot : 1-5 μg/mL, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067158
|
Novus Biologicals™
NBP3-30945-25ug |
25 μg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066902
|
Novus Biologicals™
NBP3-30945-100ug |
100 μg |
508.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CALHM1 Polyclonal antibody specifically detects CALHM1 in Human samples. It is validated for ELISA,Western BlotSpecifications
| CALHM1 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Alzheimers Research, Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| Phosphate buffered saline | |
| 255022 | |
| IgG | |
| Ammonium sulfate precipitation |
| Western Blot : 1-5 μg/mL, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| calcium homeostasis modulator 1, FAM26Ccalcium homeostasis modulator protein 1, family with sequence similarity 26, member C, MGC39514, MGC39617, Protein FAM26C | |
| Synthetic peptide derived from human CALHM1 protein. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title