missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CALHM1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30956-25ug
This item is not returnable.
View return policy
Description
CALHM1 Polyclonal antibody specifically detects CALHM1 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| CALHM1 | |
| Polyclonal | |
| Western Blot : 1:400, ELISA, Immunohistochemistry : 2-10 μg/mL | |
| calcium homeostasis modulator 1, FAM26Ccalcium homeostasis modulator protein 1, family with sequence similarity 26, member C, MGC39514, MGC39617, Protein FAM26C | |
| Synthetic peptide derived from human CALHM1 protein. | |
| 25 μg | |
| Alzheimers Research, Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 255022 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction