Learn More
BINDER™ Calibration Certificate 27 P Temperature and 1 P Humidity for Series KBF-S, KBF-S ECO
Product Code. 17323663 Shop All BINDER Products
Click to view available options
Duration:
60 to 120 min.
Unit Size:
Each
Product Code. 17323663
Brand: BINDER™ 80122070
BINDER™ Calibration certificate is an option for use with Constant Climate Chambers KBF-S and KBF-S ECO
  • Calibration certificate, temperature and humidity
  • Temperature (according to DIN12880) and humidity measurement
  • Includes certificate, 27 temperature measuring points and 1 humidity measuring point, at 25 °C / 60 % RH or at specified values
Specifications
For Use With (Equipment) BINDER Chambers Series KBF-S ECO
Description Calibration certificate, temperature and humidity - temperature (according to DIN12880) and humidity measurement incl. certificate, 27 temperature measuring points and 1 humidity measuring point, at 25°C / 60 % RH or (at specified values)
Duration 60 to 120 min.
Product Type Calibration Certificate
Includes 27 temperature measuring points and 1 humidity measuring point (at 25°C / 60 % RH or (at specified values))