Learn More
BINDER™ Calibration Certificate 9 P Temperature for Series KBF-S, KBF-S ECO
Shop All BINDER Products
Click to view available options
Duration:
60 to 120 min.
Unit Size:
Each
Description
- Calibration certificate, temperature
- Temperature measurement incl. certificate, 9 measuring points at specified temperature
Specifications
Specifications
| For Use With (Equipment) | BINDER Chambers Series KBF-S ECO |
| Description | Calibration certificate, temperature - temperature measurement incl. certificate, 9 measuring points (at specified temperature) |
| Duration | 60 to 120 min. |
| Product Type | Calibration Certificate |
| Includes | Certificate 9 measuring points (at specified temperature) |