missing translation for 'onlineSavingsMsg'
Learn More

California Oxygenates Mixture 984 Various Concentrations μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30324671
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30324671 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324671 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000984ME
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diisopropyl ether∣Ethyl tert-Butyl Ether (ETBE)∣Methyl tert-butyl ether∣tert.-Butanol∣tert-Amyl Methyl Ether (TAME)
CAS 108-20-3, 637-92-3, 1634-04-4, 75-65-0, 994-05-8
Analyte or Component Names Diisopropyl ether∣Ethyl tert-Butyl Ether (ETBE)∣Methyl tert-butyl ether∣tert.-Butanol∣tert-Amyl Methyl Ether (TAME)
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Diisopropyl ether 2000 μg/mL∣Ethyl tert-Butyl Ether (ETBE) 2000 μg/mL∣Methyl tert-butyl ether 2000 μg/mL∣tert-Amyl Methyl Ether (TAME) 2000 μg/mL∣tert.-Butanol 10000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.