missing translation for 'onlineSavingsMsg'
Learn More

California Pesticide Class 1 Mixture cis and trans Chlordane 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30309290
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30309290 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30309290 Supplier Dr. Ehrenstorfer Supplier No. DRES50000078AL
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aldicarb∣Carbofuran∣Chlorfenapyr∣Chlorpyrifos∣Cis-Chlordane (Alpha Isomer)∣Coumaphos∣Daminozide∣Dichlorvos∣Dimethoate∣Ethoprophos∣Etofenprox∣Fenoxycarb∣Fipronil∣Imazalil∣Methiocarb∣Mevinphos∣Paclobutrazol∣Parathion-methyl∣Propoxur∣Spiroxamine∣Thiacloprid∣Trans-Chlordane (Gamma Isomer)
CAS 116-06-3, 1563-66-2, 122453-73-0, 2921-88-2, 5103-71-9, 56-72-4
Analyte or Component Names Aldicarb∣Carbofuran∣Chlorfenapyr∣Chlorpyrifos∣Cis-Chlordane (Alpha Isomer)∣Coumaphos∣Daminozide∣Dichlorvos∣Dimethoate∣Ethoprophos∣Etofenprox∣Fenoxycarb∣Fipronil∣Imazalil∣Methiocarb∣Mevinphos∣Paclobutrazol∣Parathion-methyl∣Propoxur∣Spiroxamine∣Thiacloprid∣Trans-Chlordane (Gamma Isomer)
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Industry Type Food Testing
Quantity 5 x 1 mL
Concentration or Composition (by Analyte or Components) Aldicarb 100 μg/mL∣Carbofuran 100 μg/mL∣Chlorfenapyr 100 μg/mL∣Chlorpyrifos 100 μg/mL∣Cis-Chlordane (Alpha Isomer) 100 μg/mL∣Coumaphos 100 μg/mL∣Daminozide 100 μg/mL∣Dichlorvos 100 μg/mL∣Dimethoate 100 μg/mL∣Ethoprophos 100 μg/mL∣Etofenprox 100 μg/mL∣Fenoxycarb 100 μg/mL∣Fipronil 100 μg/mL∣Imazalil 100 μg/mL∣Methiocarb 100 μg/mL∣Mevinphos 100 μg/mL∣Paclobutrazol 100 μg/mL∣Parathion-methyl 100 μg/mL∣Propoxur 100 μg/mL∣Spiroxamine 100 μg/mL∣Thiacloprid 100 μg/mL∣Trans-Chlordane (Gamma Isomer) 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.