missing translation for 'onlineSavingsMsg'
Learn More
Learn More
California Pesticide Class 2B Mixture 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Hexythiazox∣Imidacloprid∣Kresoxim-methyl∣Malathion∣Metalaxyl∣Methomyl∣Myclobutanil∣Naled∣Oxamyl∣Permethrin∣Phosmet∣Piperonyl butoxide∣Prallethrin∣Propiconazole∣Pyrethrins∣Pyridaben∣Quintozene∣Spinetoram J∣Spinosad∣Spiromesifen∣Spirotetramat∣Tebuconazole∣Thiamethoxam∣Trifloxystrobin |
| CAS | 78587-05-0, 138261-41-3, 143390-89-0, 121-75-5, 57837-19-1, 16752-77-5 |
| Analyte or Component Names | Hexythiazox∣Imidacloprid∣Kresoxim-methyl∣Malathion∣Metalaxyl∣Methomyl∣Myclobutanil∣Naled∣Oxamyl∣Permethrin∣Phosmet∣Piperonyl butoxide∣Prallethrin∣Propiconazole∣Pyrethrins∣Pyridaben∣Quintozene∣Spinetoram J∣Spinosad∣Spiromesifen∣Spirotetramat∣Tebuconazole∣Thiamethoxam∣Trifloxystrobin |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetonitrile |
| Quantity | 5 x 1 mL |
| Concentration or Composition (by Analyte or Components) | Hexythiazox 100 μg/mL∣Imidacloprid 100 μg/mL∣Kresoxim-methyl 100 μg/mL∣Malathion 100 μg/mL∣Metalaxyl 100 μg/mL∣Methomyl 100 μg/mL∣Myclobutanil 100 μg/mL∣Naled 100 μg/mL∣Oxamyl 100 μg/mL∣Permethrin 100 μg/mL∣Phosmet 100 μg/mL∣Piperonyl butoxide 100 μg/mL∣Prallethrin 100 μg/mL∣Propiconazole 100 μg/mL∣Pyrethrins 100 μg/mL∣Pyridaben 100 μg/mL∣Quintozene 100 μg/mL∣Spinetoram J 100 μg/mL∣Spinosad 100 μg/mL∣Spiromesifen 100 μg/mL∣Spirotetramat 100 μg/mL∣Tebuconazole 100 μg/mL∣Thiamethoxam 100 μg/mL∣Trifloxystrobin 100 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Haben Sie Verbesserungsvorschläge?