missing translation for 'onlineSavingsMsg'
Learn More
Learn More
California Pesticides Class 2A Mixture 1034 100 μg/mL in Acetonitrile, Second Source, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Abamectin∣Acephate∣Acequinocyl∣Acetamiprid∣Azoxystrobin∣Bifenazate∣Bifenthrin∣Boscalid∣Captan∣Carbaryl∣Chlorantraniliprole∣Clofentezine∣Cyfluthrin∣Cypermethrin∣Diazinon∣Dimethomorph∣Etoxazole∣Fenhexamid∣Fenpyroximate (E/Z Mix)∣Flonicamid∣Fludioxonil |
| CAS | 71751-41-2, 30560-19-1, 57960-19-7, 135410-20-7, 131860-33-8, 149877-41-8 |
| Analyte or Component Names | Abamectin∣Acephate∣Acequinocyl∣Acetamiprid∣Azoxystrobin∣Bifenazate∣Bifenthrin∣Boscalid∣Captan∣Carbaryl∣Chlorantraniliprole∣Clofentezine∣Cyfluthrin∣Cypermethrin∣Diazinon∣Dimethomorph∣Etoxazole∣Fenhexamid∣Fenpyroximate (E/Z Mix)∣Flonicamid∣Fludioxonil |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetonitrile |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Abamectin 100 μg/mL∣Acephate 100 μg/mL∣Acequinocyl 100 μg/mL∣Acetamiprid 100 μg/mL∣Azoxystrobin 100 μg/mL∣Bifenazate 100 μg/mL∣Bifenthrin 100 μg/mL∣Boscalid 100 μg/mL∣Captan 100 μg/mL∣Carbaryl 100 μg/mL∣Chlorantraniliprole 100 μg/mL∣Clofentezine 100 μg/mL∣Cyfluthrin 100 μg/mL∣Cypermethrin 100 μg/mL∣Diazinon 100 μg/mL∣Dimethomorph 100 μg/mL∣Etoxazole 100 μg/mL∣Fenhexamid 100 μg/mL∣Fenpyroximate (E/Z Mix) 100 μg/mL∣Flonicamid 100 μg/mL∣Fludioxonil 100 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?