missing translation for 'onlineSavingsMsg'
Learn More

California Pesticides Class 2A Mixture 668 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30323145
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30323145 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30323145 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000668AL
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material Abamectin∣Acephate∣Acequinocyl∣Acetamiprid∣Azoxystrobin∣Bifenazate∣Bifenthrin∣Boscalid∣Captan∣Carbaryl∣Chlorantraniliprole∣Clofentezine∣Cyfluthrin∣Cypermethrin∣Diazinon∣Dimethomorph∣Etoxazole∣Fenhexamid∣Fenpyroximate (E/Z Mix)∣Flonicamid∣Fludioxonil
CAS 71751-41-2, 30560-19-1, 57960-19-7, 135410-20-7, 131860-33-8, 149877-41-8
Analyte or Component Names Abamectin∣Acephate∣Acequinocyl∣Acetamiprid∣Azoxystrobin∣Bifenazate∣Bifenthrin∣Boscalid∣Captan∣Carbaryl∣Chlorantraniliprole∣Clofentezine∣Cyfluthrin∣Cypermethrin∣Diazinon∣Dimethomorph∣Etoxazole∣Fenhexamid∣Fenpyroximate (E/Z Mix)∣Flonicamid∣Fludioxonil
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Abamectin 100 μg/mL∣Acephate 100 μg/mL∣Acequinocyl 100 μg/mL∣Acetamiprid 100 μg/mL∣Azoxystrobin 100 μg/mL∣Bifenazate 100 μg/mL∣Bifenthrin 100 μg/mL∣Boscalid 100 μg/mL∣Captan 100 μg/mL∣Carbaryl 100 μg/mL∣Chlorantraniliprole 100 μg/mL∣Clofentezine 100 μg/mL∣Cyfluthrin 100 μg/mL∣Cypermethrin 100 μg/mL∣Diazinon 100 μg/mL∣Dimethomorph 100 μg/mL∣Etoxazole 100 μg/mL∣Fenhexamid 100 μg/mL∣Fenpyroximate (E/Z Mix) 100 μg/mL∣Flonicamid 100 μg/mL∣Fludioxonil 100 μg/mL
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.