missing translation for 'onlineSavingsMsg'
Learn More

Calix[4]arene, 98%

Product Code. 10712004 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10712004 5 g Glass bottle 5g
10165210 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10712004 Supplier Thermo Scientific Acros Supplier No. 290190050
 Restricted Item
Estimated Shipment: 08-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 74568-07-3 | C28H24O4 | 424.5 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74568-07-3
Molecular Formula C28H24O4
Molecular Weight (g/mol) 424.5
MDL Number MFCD00233673
InChI Key YPNHVQZZPXPQOS-UHFFFAOYSA-N
Synonym calix 4 arene, tetrahydroxycalix 4 arene, calix 4 arene 1g, calix 4 arene-25,26,27,28-tetrol, 25,26,27,28-tetrahydroxycalix 4 aren, 25,26,27,28-tetrahydroxycalix 4 arene, calix 4 arene-25,26,27,28-tetrol, purum hplc
PubChem CID 562409
SMILES C1C2=C(C(=CC=C2)CC3=C(C(=CC=C3)CC4=CC=CC(=C4O)CC5=CC=CC1=C5O)O)O

Specifications

Melting Point 315.0°C to 318.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 424.5
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Calix[4]arene, 98%
compliance-icons
Product Identifier
  • Calix[4]arene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.