missing translation for 'onlineSavingsMsg'
Learn More

Calix[4]arene, 98%

Product Code. 11499227
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499227 1 g 1g
11409237 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499227 Supplier Thermo Scientific Alfa Aesar Supplier No. B21320.03
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 74568-07-3 | C28H24O4 | 424.496 g/mol

Functionalized calix[4]arenes can bind DNA and induce cell transfection. Calixarenes lend themselves well to many applications because of the multiplicity of options for such structural elaboration such as in electrochemical sensors, optical sensors, chiral recognition devices,

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Functionalized calix[4]arenes can bind DNA and induce cell transfection. Calixarenes lend themselves well to many applications because of the multiplicity of options for such structural elaboration such as in electrochemical sensors, optical sensors, chiral recognition devices,

Solubility
Soluble in chloroform.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74568-07-3
Molecular Formula C28H24O4
Molecular Weight (g/mol) 424.496
MDL Number MFCD00143912
InChI Key YPNHVQZZPXPQOS-UHFFFAOYSA-N
Synonym calix 4 arene, tetrahydroxycalix 4 arene, calix 4 arene 1g, calix 4 arene-25,26,27,28-tetrol, 25,26,27,28-tetrahydroxycalix 4 aren, 25,26,27,28-tetrahydroxycalix 4 arene, calix 4 arene-25,26,27,28-tetrol, purum hplc
PubChem CID 562409
SMILES C1C2=C(C(=CC=C2)CC3=C(C(=CC=C3)CC4=CC=CC(=C4O)CC5=CC=CC1=C5O)O)O

Specifications

Melting Point 315°C to 318°C
Quantity 1 g
Solubility Information Soluble in chloroform.
Formula Weight 424.5
Percent Purity 98%
Chemical Name or Material Calix[4]arene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.