missing translation for 'onlineSavingsMsg'
Learn More

Calix[6]arene, 98%

Product Code. 11497224
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497224 25 g 25g
11477224 1 g 1g
11487224 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11497224 Supplier Thermo Scientific Alfa Aesar Supplier No. B21608.14
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 96107-95-8 | C42H36O6 | 636.744 g/mol

It is used as a starting material for preparing functionalized calixarenes. It finds its uses as a pharmaceutical intermediate. It is also used in complexation and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a starting material for preparing functionalized calixarenes. It finds its uses as a pharmaceutical intermediate. It is also used in complexation and organic synthesis.

Solubility
0.180g/100g of water @ 25°C. Miscible with alcohol, chloroform, ether, carbon disulfide, acetone, oils, carbon tetrachloride and glacial acetic acid.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96107-95-8
Molecular Formula C42H36O6
Molecular Weight (g/mol) 636.744
MDL Number MFCD00143083
InChI Key JLSWUKWFQCVKCL-UHFFFAOYSA-N
Synonym calix 6 arene, hexahydroxycalix 6 arene
PubChem CID 2724885
SMILES C1C2=C(C(=CC=C2)CC3=C(C(=CC=C3)CC4=C(C(=CC=C4)CC5=CC=CC(=C5O)CC6=CC=CC(=C6O)CC7=CC=CC1=C7O)O)O)O

Specifications

Melting Point >300°C
Quantity 25 g
Solubility Information 0.180g/100g of water @ 25°C. Miscible with alcohol,chloroform,ether,carbon disulfide,acetone,oils,carbon tetrachloride and glacial acetic acid.
Formula Weight 636.74
Percent Purity 98%
Chemical Name or Material Calix[6]arene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.