missing translation for 'onlineSavingsMsg'
Learn More

Calix[8]arene, 97%

Product Code. 11490237
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490237 1 g 1g
11400247 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11490237 Supplier Thermo Scientific Alfa Aesar Supplier No. B21666.03
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 82452-93-5 | C56H48O8 | 848.992 g/mol

It is used in the study on the chromatographic behavior of water-soluble vitamins on p-tert-butyl-calix [8] arene-bonded silica gel stationary phase by HPLC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the study on the chromatographic behavior of water-soluble vitamins on p-tert-butyl-calix [8] arene-bonded silica gel stationary phase by HPLC.

Solubility
Sparingly soluble in water (5.8E-6 g/L) (25°C).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82452-93-5
Molecular Formula C56H48O8
Molecular Weight (g/mol) 848.992
MDL Number MFCD00143914
InChI Key HDPRHRZFFPXZIL-UHFFFAOYSA-N
Synonym calix 8 arene, calix 8 arene;, p-h-calix 8 arene, octahydroxycalix 8 arene, para-h-calix 8 arene;, calix 8 arene, technical ch, 49,50,51,52,53,54,55,56-octahydroxycalix 8 arene, 49,50,51,52,53,54,55,56-octahydroxycalix 8 arene;, 49,50,51,52,53,54,55,56-octahydroxycalix 8-arene;, 49,50,51,52,53,54,55,56-octakis hydroxy calix 8 arene;
PubChem CID 2724886
SMILES C1C2=C(C(=CC=C2)CC3=C(C(=CC=C3)CC4=C(C(=CC=C4)CC5=C(C(=CC=C5)CC6=CC=CC(=C6O)CC7=CC=CC(=C7O)CC8=CC=CC(=C8O)CC9=CC=CC1=C9O)O)O)O)O

Specifications

Melting Point >450°C
Quantity 1 g
Solubility Information Sparingly soluble in water (5.8E-6g/L) (25°C).
Formula Weight 848.99
Percent Purity 97%
Chemical Name or Material Calix[8]arene
compliance-icons
Product Identifier
  • Calix(8)arene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Calix(8)arene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.