Learn More
Calnexin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 11 publications
Brand:  Novus Biologicals NBP1-97485-0.025MG
 View more versions of this product
Product Code. 18049537
  • 193.00 € / 25µg
Description
Calnexin Polyclonal specifically detects Calnexin in Human, Mouse, Rat, Porcine, Avian, Bovine, Canine, Chicken, Drosophila, Guinea Pig, Hamster, Primate, Rabbit, Sheep, Xenopus samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Electron Microscopy.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Calnexin
Polyclonal
Western Blot 1:1000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Electron Microscopy 1:10-1:500
P24643
CANX
Synthetic peptide corresponding to the sequence near the C-terminus of canine calnexin.
0.025 mg
Primary
Pig, Chicken, Rat, Canine, Avian, Mouse, Drosophila, Human, Sheep, Primate, Guinea Pig, Rabbit, Hamster, Bovine
Purified
Flow Cytometry, Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry, Immunocytochemistry
Unconjugated
PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide
calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90
Rabbit
Protein A purified
RUO
821
Store at -20C. Avoid freeze-thaw cycles.