missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Calnexin Monoclonal Antibody (AF18)

Product Code. 11555742
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11555742 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 11555742 Supplier Invitrogen™ Supplier No. MA3027
Keep Your Science Moving with 3 for 2
Use promo code 27970
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price

Mouse Monoclonal Antibody

MA3-027 detects calnexin from human and mouse tissues. MA3-027 has been successfully used in Western blot, immunocytochemistry, immunofluorescence and immunoprecipitation protocols. By Western blot, this antibody detects calnexin in human liver, skin, adrenal gland, heart, colon, testes and ovary extracts. This antibody is not recommended for human kidney or mouse liver lysates in Western blot applications. The predicted molecular weight of this antibody is ~67kDa; however, it may run on SDS-PAGE as an ~90kDA protein. This is due mainly to negative charges at the C-terminus of the protein, which effects the SDS binding to the molecules and consequently its SDS-PAGE mobility. Immunocytochemical staining of calnexin in Huh-7 cells with MA3-027 yields a pattern consistent with specific endoplasmic reticulum staining. The MA3-027 antigen is Focus human hepatoma cell lysate.

Calnexin, also referred to as IP90, p88 and p90, is an approximately 90 kDa integral membrane protein of the endoplasmic reticulum (ER). Many resident ER proteins act as molecular chaperones and participate in the proper folding of polypeptides and their assembly into multisubunit proteins. Studies indicate that calnexin associates with the major histocompatibility complex (MHC) class I heavy chains, partial complexes of the T cell receptor and B cell membrane immunoglobulin, but not with completed receptor complexes. It has been shown that calnexin is a chaperone that retains incompletely or improperly folded proteins in the ER. The sequence Lys-Asp-Glu-Leu (KDEL) or a closely related sequence, is present at the carboxy-terminus of soluble ER resident proteins such as GRP78 and GRP94 and protein disulfide isomerase. Integral membrane ER resident proteins, like calnexin, often lack this KDEL sequence but contain positively charged cytosolic residues that ensure ER retention. Calnexin contains a large ER luminal domain (461 amino acids), a transmembrane segment (22 amino acids), and a cytoplasmic tail (89 amino acids).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calnexin
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, RNA Immunoprecipitation, Immunocytochemistry
Classification Monoclonal
Clone AF18
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene CANX
Gene Accession No. P27824, P35564
Gene Alias 1110069N15Rik; AI988026; At5g07340; At5g61790; calmegin; calnexin; canx; canx-a; canx-b; clgn; clnx; cnx; CNX1; CNX2; D11Ertd153e; FLJ26570; hypothetical protein; IP90; MAC9.11; Major histocompatibility complex class I antigen-binding protein p88; p88; P90; PP90; pp90 precursor
Gene Symbols CANX
Host Species Mouse
Immunogen Focus human hepatoma cell lysate.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12330, 821
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.