missing translation for 'onlineSavingsMsg'
Learn More
Learn More
calreticulin Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
129.00 € - 399.00 €
Specifications
| Antigen | calreticulin |
|---|---|
| Clone | 2B17 |
| Concentration | 350 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236175
|
Proteintech
80017-1-RR-100UL |
100 μL |
439.00 € 399.00 € / 100µL Save 40.00 € 9% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234063
|
Proteintech
80017-1-RR-20UL |
20 μL |
149.00 € 129.00 € / 20µL Save 20.00 € 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The calreticulin antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human calreticulin. This antibody recognizes human, mouse, rat antigen. The calreticulin antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| calreticulin | |
| 350 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| 811 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 2B17 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| CRT; FLJ26680; RO; SSA; cC1qR | |
| IgG | |
| Protein A purification | |
| Recombinant Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title