missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calretinin Antibody (3G9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50042SS
This item is not returnable.
View return policy
Description
Calretinin Monoclonal antibody specifically detects Calretinin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry Free-Floating
Specifications
| Calretinin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:1000, Immunocytochemistry/ Immunofluorescence 1:1000, Immunohistochemistry Free-Floating | |
| P22676 | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Free Floating) | |
| 3G9 | |
| Unconjugated | |
| 50% PBS, 50% glycerol | |
| 29 kDa calbindin, CAB29, CAL2, calbindin 2, calbindin 2, (29kD, calretinin), calbindin 2, 29kDa (calretinin), calbindin D29K, calretinin, CR | |
| Full-length recombinant human Calretinin expressed in and purified from E. coli. [UniProt# P22676] | |
| 0.025 mL | |
| Cellular Markers | |
| 794 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction