missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calretinin Antibody (CL12836), Novus Biologicals™
Mouse Monoclonal Antibody
389.00 € - 580.00 €
Specifications
| Antigen | Calretinin |
|---|---|
| Clone | CL12836 |
| Dilution | Western Blot 1 μg/ml, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18636154
|
Novus Biologicals
NBP3-18575-100ul |
100 μg |
580.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18637394
|
Novus Biologicals
NBP3-18575-25ul |
25 μg |
389.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calretinin Monoclonal antibody specifically detects Calretinin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Calretinin | |
| Western Blot 1 μg/ml, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| 29 kDa calbindin, CAB29, CAL2, calbindin 2, calbindin 2, (29kD, calretinin), calbindin 2, 29kDa (calretinin), calbindin D29K, calretinin, CR | |
| This antibody was developed against a recombinant protein of P22676. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL12836 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cellular Markers, Phospho Specific | |
| PBS, pH 7.2, 40% glycerol | |
| 794 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title