missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CaM Kinase II gamma Antibody (6A4A10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37269
This item is not returnable.
View return policy
Description
CaM Kinase II gamma Monoclonal specifically detects CaM Kinase II gamma in Human samples. It is validated for Western Blot, ELISA.
Specifications
| CaM Kinase II gamma | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| calcium/calmodulin-dependent protein kinase (CaM kinase) II gamma, calcium/calmodulin-dependent protein kinase II gamma, calcium/calmodulin-dependent protein kinase type II subunit gamma, CaM kinase II subunit gamma, CAMK, CAMKGFLJ16043, CAMK-II, CaMK-II subunit gamma, EC 2.7.11, EC 2.7.11.17, MGC26678 | |
| Mouse | |
| 62.6 kDa | |
| 0.1 mL | |
| Protein Kinase, Signal Transduction, Tyrosine Kinases, Wnt Signaling Pathway | |
| 818 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 6A4A10 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| CAMK2G | |
| Purified recombinant fragment of human CAMK2G (AA: 322-481) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction