missing translation for 'onlineSavingsMsg'
Learn More

CaMKII alpha/beta, p Thr286, p Thr287 Antibody (22B1), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 4 publications

Brand:  Novus Biologicals NB110-96869-0.025mg

 View more versions of this product

Product Code. 18575942

  • 191.00 € / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CaMKII alpha/beta Monoclonal specifically detects CaMKII alpha/beta in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CaMKII alpha/beta (p Thr286, p Thr287)
Monoclonal
1 mg/mL
Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunoblotting
calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha, calcium/calmodulin-dependent protein kinase II alpha, calcium/calmodulin-dependent protein kinase II alpha-B subunit, CaM kinase II alpha subunit, CaM kinase II subunit alpha, CAMKAcalcium/calmodulin-dependent protein kinase type II subunit alpha, CaMK-II alpha subunit, CaMK-II subunit alpha, CaMKIINalpha, CaM-kinase II alpha chain, EC 2.7.11, EC 2.7.11.17, KIAA0968calcium/calmodulin-dependent protein kinase type II alpha chain
Mouse
Protein G purified
RUO
Primary
This antibody is specific for a and B subunits of CaMKII only when they are phosphorylated at Thr-286/287 (in B).
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
IgG1
Western Blot
22B1
Unconjugated
P11275
CAMK2A
Synthetic peptide
0.025 mg
Phospho Specific, Wnt Signaling Pathway
815
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

For Research Use Only