missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAMKK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13624-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody recognize the N-terminal of CAMKK1 .
The product of this gene is a component of the nuclear factor of activated T cells DNA-binding transcription complex. This complex consists of at least two components: a preexisting cytosolic component that translocates to the nucleus upon T cell receptor (TCR) stimulation, and an inducible nuclear component. Proteins belonging to this family of transcription factors play a central role in inducible gene transcription during immune response. The product of this gene is an inducible nuclear component. It functions as a major molecular target for the immunosuppressive drugs such as cyclosporin A. Five transcript variants encoding distinct isoforms have been identified for this gene. Different isoforms of this protein may regulate inducible expression of different cytokine genes.Specifications
| CAMKK1 | |
| Polyclonal | |
| Unconjugated | |
| CAMKK1 | |
| CaM kinase IV kinase, CaM kinase kinase 1, CaM kinase kinase alpha, CaM KK 1, CaM KK alpha, CaMKK 1, CaMKK alpha, CAMKK1, CAMKKA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55984, 60341, 84254 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P97756, Q8N5S9, Q8VBY2 | |
| Camkk1 | |
| CAMKK1 Fusion Protein Ag4534 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction