missing translation for 'onlineSavingsMsg'
Learn More

Camostat Mesylate, TRC

Product Code. 30136252
Change view
Click to view available options
Quantity:
10 mg
1 g
2.5 g
Unit Size:
10mg
1g
2.50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136252 1 g 1g
30134353 2.5 g 2.50g
30007921 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136252 Supplier Toronto Research Chemicals Supplier No. TRCC1503001G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59721-29-8
Molecular Formula C20 H22 N4 O5 . C H4 O3 S
Molecular Weight (g/mol) 494.52
Synonym Benzeneacetic acid, 4-[[4-[(aminoiminomethyl)amino]benzoyl]oxy]-, 2-(dimethylamino)-2-oxoethyl ester, methanesulfonate (1:1), Benzeneacetic acid, 4-[[4-[(aminoiminomethyl)amino]benzoyl]oxy]-, 2-(dimethylamino)-2-oxoethyl ester, monomethanesulfonate (9CI), Camostat mesylate, Camostat monomethanesulfonate, FOY 305, FOY-S 980, Foipan
IUPAC Name [4-[2-[2-(dimethylamino)-2-oxoethoxy]-2-oxoethyl]phenyl] 4-carbamimidamidobenzoate;methanesulfonic acid
SMILES CN(C)C(=O)COC(=O)Cc1ccc(OC(=O)c2ccc(NC(=N)N)cc2)cc1.CS(=O)(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C20H22N4O5.CH4O3S/c1-24(2)17(25)12-28-18(26)11-13-3-9-16(10-4-13)29-19(27)14-5-7-15(8-6-14)23-20(21)22;1-5(2,3)4/h3-10H,11-12H2,1-2H3,(H4,21,22,23);1H3,(H,2,3,4)
Formula Weight 494.1471
Chemical Name or Material Camostat Mesylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.