missing translation for 'onlineSavingsMsg'
Learn More

Camphene 1000 μg/mL in Isopropanol, Dr. Ehrenstorfer

Product Code. 30328416
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
Product Code. Quantity unitSize
30328416 5 x 1 mL 5mL
1 options
This item is not returnable. View return policy

Product Code. 30328416

Brand: Dr. Ehrenstorfer DREGS09010073IP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Camphene
CAS 79-92-5
Analyte or Component Names Camphene
Shipping Condition Room Temperature
Molecular Formula C10 H16
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Bicyclo[2.2.1]heptane, 2,2-dimethyl-3-methylene-, Camphene (8CI), 2,2-Dimethyl-3-methylenebicyclo[2.2.1]heptane, (±)-Camphene, 2,2-Dimethyl-3-methylenenorbornane, 2-Methylene-3,3-dimethylbicyclo[2.2.1]heptane, 3,3-Dimethyl-2-methylenenorbornane, 3,3-Dimethyl-2-methylenenorcamphane, DL-Camphene, NSC 4165, YS Camphene, dl-Camphene
InChI Formula InChI=1S/C10H16/c1-7-8-4-5-9(6-8)10(7,2)3/h8-9H,1,4-6H2,2-3H3/t8-,9+/m0/s1
SMILES CC1(C)[C@@H]2CC[C@@H](C2)C1=C
IUPAC Name (1S,4R)-3,3-dimethyl-2-methylidenebicyclo[2.2.1]heptane
Solvent or Matrix Isopropanol
Molecular Weight (g/mol) 136.23
Quantity 5 x 1 mL
Formula Weight 136.1252 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.