missing translation for 'onlineSavingsMsg'
Learn More

Camphene, SPEX CertiPrep™

Product Code. 16340448 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
16340448 1 mL 1mL
1 options
This item is not returnable. View return policy

Product Code. 16340448

Brand: SPEX Certiprep S710

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
(+)-Camphene5794-03-61000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Packaging 2mL amber prescored glass ampule
CAS 79-92-5, 67-56-1
CAS Min % 0.1, 99.9
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Molecular Formula C10H16
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI Key CRPUJAZIXJMDBK-UHFFFAOYSA-N
Type Certified Reference Material
SMILES CC1(C2CCC(C2)C1=C)C
IUPAC Name 3,3-dimethyl-2-methylidenebicyclo[2.2.1]heptane
Solvent or Matrix Methanol-P and T
Molecular Weight (g/mol) 136.238
PubChem CID 6616
Quantity 1 mL
ChEBI CHEBI:3830
Chemical Name or Material Camphene
Show More Show Less

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.