missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Camphene, tech. (sum of camphene + fenchene)

Product Code. 11439186
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
Product Code. Quantity unitSize
11439186 250 g 250g
11429186 50 g 50g
11419186 1000 g 1000g
3 options
This item is not returnable. View return policy

Product Code. 11439186

Brand: Thermo Scientific Alfa Aesar A18040.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 79-92-5 | C10H16 | 136.24 g/mol

Camphene is used in the preparation of fragrances and as a food additive for flavoring.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Camphene is used in the preparation of fragrances and as a food additive for flavoring.

Solubility
Insoluble in water. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-92-5
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00066603
InChI Key CRPUJAZIXJMDBK-UHFFFAOYNA-N

Specifications

Melting Point ∼35°C
Boiling Point 159°C to 160°C
Flash Point 34°C (94°F)
Quantity 250 g
UN Number UN1325
Merck Index 14,1730
Synonym dmbi, 3-4-dimethylamino benzylidene-1,3-dihydro-2h-indol-2-one, 3z-3-4-dimethylamino phenyl methylidene-1h-indol-2-one, 3-4-dimethylamino-benzylidenyl-2-indolinone, biomolki_000054, biomolki2_000060, upcmld-dp118, bmk1-f6, upcmld-dp118:001, upcmld-dp118:002
SMILES CC1(C)C2CCC(C2)C1=C
IUPAC Name (3Z)-3-[[4-(dimethylamino)phenyl]methylidene]-1H-indol-2-one
PubChem CID 6450842
Formula Weight 136.24
Grade Technical
Chemical Name or Material (+/-)-Camphene, sum of camphene + fenchene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.