accessibility menu, dialog, popup

UserName

CAS RN 79-92-5

H3CH3CH2C

CAS RN 79-92-5

IUPAC Name: 2,2-dimethyl-3-methylidenebicyclo[2.2.1]heptane
Synonyms: camphene L-camphene comphene
Molecular Weight (g/mol): 136.24
Molecular Formula: C10H16
InChi Key: CRPUJAZIXJMDBK-UHFFFAOYNA-N
SMILES: CC1(C)C2CCC(C2)C1=C
Molecular Weight (g/mol) 
  • (5)
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Grade 
  • (3)
Solvent or Matrix 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (5)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)