missing translation for 'onlineSavingsMsg'
Learn More

Canada Pesticide Mixture 1 50 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30311018
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30311018 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30311018 Supplier Dr. Ehrenstorfer Supplier No. DREGA09001041AL
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Abamectin∣Acetamiprid∣Aldicarb∣Bifenazate∣Boscalid∣Carbofuran∣Chlorantraniliprole∣Daminozide∣Diazinon∣Dichlorvos∣Dimethoate∣Dinotefuran∣Ethoprophos∣Fenpyroximate (E/Z Mix)∣Flonicamid∣Imidacloprid∣Malathion∣Metalaxyl∣Methiocarb∣Methomyl∣Myclobutanil∣Novaluron∣Oxamyl∣Paclobutrazol∣Phosmet∣Piperonyl butoxide∣Propoxur∣Spinetoram J∣Spinosad∣Spiromesifen∣Spirotetramat∣Tebuconazole∣Thiacloprid∣Thiamethoxam∣Thiophanate-methyl
CAS 71751-41-2, 135410-20-7, 116-06-3, 149877-41-8, 188425-85-6, 1563-66-2
Analyte or Component Names Abamectin∣Acetamiprid∣Aldicarb∣Bifenazate∣Boscalid∣Carbofuran∣Chlorantraniliprole∣Daminozide∣Diazinon∣Dichlorvos∣Dimethoate∣Dinotefuran∣Ethoprophos∣Fenpyroximate (E/Z Mix)∣Flonicamid∣Imidacloprid∣Malathion∣Metalaxyl∣Methiocarb∣Methomyl∣Myclobutanil∣Novaluron∣Oxamyl∣Paclobutrazol∣Phosmet∣Piperonyl butoxide∣Propoxur∣Spinetoram J∣Spinosad∣Spiromesifen∣Spirotetramat∣Tebuconazole∣Thiacloprid∣Thiamethoxam∣Thiophanate-methyl
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Abamectin 50 μg/mL∣Acetamiprid 50 μg/mL∣Aldicarb 50 μg/mL∣Bifenazate 50 μg/mL∣Boscalid 50 μg/mL∣Carbofuran 50 μg/mL∣Chlorantraniliprole 50 μg/mL∣Daminozide 50 μg/mL∣Diazinon 50 μg/mL∣Dichlorvos 50 μg/mL∣Dimethoate 50 μg/mL∣Dinotefuran 50 μg/mL∣Ethoprophos 50 μg/mL∣Fenpyroximate (E/Z Mix) 50 μg/mL∣Flonicamid 50 μg/mL∣Imidacloprid 50 μg/mL∣Malathion 50 μg/mL∣Metalaxyl 50 μg/mL∣Methiocarb 50 μg/mL∣Methomyl 50 μg/mL∣Myclobutanil 50 μg/mL∣Novaluron 50 μg/mL∣Oxamyl 50 μg/mL∣Paclobutrazol 50 μg/mL∣Phosmet 50 μg/mL∣Piperonyl butoxide 50 μg/mL∣Propoxur 50 μg/mL∣Spinetoram J 50 μg/mL∣Spinosad 50 μg/mL∣Spiromesifen 50 μg/mL∣Spirotetramat 50 μg/mL∣Tebuconazole 50 μg/mL∣Thiacloprid 50 μg/mL∣Thiamethoxam 50 μg/mL∣Thiophanate-methyl 50 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.