missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canada Pesticide Mixture 2 ver. 2 20-1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Abamectin∣Acephate∣Allethrin∣Bifenthrin∣Chlorpyrifos∣Clofentezine∣Coumaphos∣Cypermethrin∣Diazinon∣Dichlorvos∣Dimethoate∣Ethoprophos∣Fensulfothion∣Fenthion∣Fipronil∣Kresoxim-methyl∣Metalaxyl∣Methomyl∣Paclobutrazol∣Phenothrin∣Prallethrin∣Propiconazole∣Propoxur∣Pyraclostrobin∣Pyridaben∣Resmethrin∣Spirotetramat∣Teflubenzuron∣Tetramethrin |
| CAS | 71751-41-2, 30560-19-1, 584-79-2, 82657-04-3, 2921-88-2, 74115-24-5 |
| Analyte or Component Names | Abamectin∣Acephate∣Allethrin∣Bifenthrin∣Chlorpyrifos∣Clofentezine∣Coumaphos∣Cypermethrin∣Diazinon∣Dichlorvos∣Dimethoate∣Ethoprophos∣Fensulfothion∣Fenthion∣Fipronil∣Kresoxim-methyl∣Metalaxyl∣Methomyl∣Paclobutrazol∣Phenothrin∣Prallethrin∣Propiconazole∣Propoxur∣Pyraclostrobin∣Pyridaben∣Resmethrin∣Spirotetramat∣Teflubenzuron∣Tetramethrin |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetonitrile |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Acephate 19.7 μg/mL∣Paclobutrazol 19.72 μg/mL∣Fensulfothion 19.75 μg/mL∣Metalaxyl 19.75 μg/mL∣Spirotetramat 19.76 μg/mL∣Ethoprophos 19.79 μg/mL∣Kresoxim-methyl 19.81 μg/mL∣Dimethoate 19.82 μg/mL∣Fenthion 19.84 μg/mL∣Clofentezine 19.85 μg/mL∣Pyraclostrobin 19.85 μg/mL∣Coumaphos 19.9 μg/mL∣Propoxur 19.93 μg/mL∣Diazinon 20.21 μg/mL∣Chlorpyrifos 39.86 μg/mL∣Teflubenzuron 49.69 μg/mL∣Phenothrin 49.84 μg/mL∣Pyridaben 49.87 μg/mL∣Prallethrin 50.08 μg/mL∣Methomyl 50.22 μg/mL∣Fipronil 59.71 μg/mL∣Propiconazole 98.43 μg/mL∣Resmethrin 99.25 μg/mL∣Tetramethrin 99.74 μg/mL∣Abamectin 100.1 μg/mL∣Dichlorvos 102 μg/mL∣Allethrin 198.7 μg/mL∣Cypermethrin 296.1 μg/mL∣Bifenthrin 992.2 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?