missing translation for 'onlineSavingsMsg'
Learn More

Candesartan, 98%

Product Code. 15405129
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15405129 100 mg 100mg
15495119 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15405129 Supplier Thermo Scientific Alfa Aesar Supplier No. J62818.MC
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Angiotensin II receptor antagonist | CAS: 139481-59-7 | C24H20N6O3 | 440.463 g/mol

Candesartan is a selective AT1 (angiotensin II receptor 1) antagonist. Antagonism of angiotensin receptors inhibits vasoconstriction and the production of aldosterone, leading to a decrease in water and sodium concentration in blood plasma. Exhibits antihypertensive effects in animal models. Used in treatment of congestive heart failure, as antihypertensive. Candesartan does not affect cell viability or proliferation but increases the expression of VEGF and interleukin-8 in the cultured medium of KU-19-19 cells. Candesartan (0.1 nM) could reduce the maximal contractile response to angiostensin II by approximately 50%.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in ethyl acetate, methanol, water (<1mg/mL at 25°C), DMSO (88 mg/ml at 25°C), and ethanol (1 mg/ml at 25°C).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139481-59-7
Molecular Formula C24H20N6O3
Molecular Weight (g/mol) 440.463
MDL Number MFCD00081076
InChI Key HTQMVQVXFRQIKW-UHFFFAOYSA-N
Synonym candesartan, blopress, ratacand, 1-2'-1h-tetrazol-5-yl-1,1'-biphenyl-4-yl methyl-2-ethoxy-1h-benzo d imidazole-7-carboxylic acid, candesartan ban, 3h candesartan, unii-s8q36md2xx, 2-ethoxy-3-4-2-2h-tetrazol-5-yl phenyl phenyl methyl benzimidazole-4-carboxylic acid, candesartan usan/inn, candesartan usan:inn
PubChem CID 2541
ChEBI CHEBI:3347
IUPAC Name 2-ethoxy-3-[[4-[2-(2H-tetrazol-5-yl)phenyl]phenyl]methyl]benzimidazole-4-carboxylic acid
SMILES CCOC1=NC2=CC=CC(=C2N1CC3=CC=C(C=C3)C4=CC=CC=C4C5=NNN=N5)C(=O)O

Specifications

Melting Point 183°C to 185°C
Color White
Quantity 100 mg
Merck Index 14,1739
Solubility Information Soluble in ethyl acetate,methanol,water (<1mg/ml at 25°C),DMSO (88mg/ml at 25°C),and ethanol (1mg/ml at 25°C).
Formula Weight 440.45
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Candesartan
compliance-icons
Product Identifier
  • Candesartan
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361d-Suspected of damaging the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Candesartan

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.