accessibility menu, dialog, popup

UserName

H3CONHOONNHNNN

CAS RN 139481-59-7

IUPAC Name: 2-ethoxy-1-{[2'-(2H-1,2,3,4-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl]methyl}-1H-1,3-benzodiazole-7-carboxylic acid
Synonyms: candesartan 坎地沙坦 ratacand amias candesartan cilexetil atacand
Molecular Weight (g/mol): 440.46
Molecular Formula: C24H20N6O3
InChi Key: HTQMVQVXFRQIKW-UHFFFAOYSA-N
SMILES: CCOC1=NC2=CC=CC(C(O)=O)=C2N1CC1=CC=C(C=C1)C1=CC=CC=C1C1=NNN=N1
Molecular Weight (g/mol) 
  • (8)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Certifications/Compliance 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (8)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)