Learn More
Candesartan, 98%
Description
Candesartan is a selective AT1 (angiotensin II receptor 1) antagonist. Antagonism of angiotensin receptors inhibits vasoconstriction and the production of aldosterone, leading to a decrease in water and sodium concentration in blood plasma. Exhibits antihypertensive effects in animal models. Used in treatment of congestive heart failure, as antihypertensive. Candesartan does not affect cell viability or proliferation but increases the expression of VEGF and interleukin-8 in the cultured medium of KU-19-19 cells. Candesartan (0.1 nM) could reduce the maximal contractile response to angiostensin II by approximately 50%.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in ethyl acetate, methanol, water (<1mg/mL at 25°C), DMSO (88 mg/ml at 25°C), and ethanol (1 mg/ml at 25°C).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 183°C to 185°C |
| Color | White |
| Quantity | 1 g |
| Merck Index | 14,1739 |
| Solubility Information | Soluble in ethyl acetate,methanol,water (<1mg/ml at 25°C),DMSO (88mg/ml at 25°C),and ethanol (1mg/ml at 25°C). |
| Formula Weight | 440.45 |
| Percent Purity | 98% |
| Physical Form | Powder |
| Chemical Name or Material | Candesartan |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.