missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Candesartan
GREENER_CHOICE

Product Code. 30001557
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001557 10 mg 10mg
30001575 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001557 Supplier Tocris Bioscience™ Supplier No. 4791/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

AT1 receptor antagonist

Angiotensin II receptor 1 (AT1) antagonist (IC50 values are 1.12 and 2.86 nM in bovine adrenal cortex and rabbit aorta, respectively). Exhibits antihypertensive effects in animal models. Also available as a prodrug, candesartan cilexetil (Cat. No. 4792).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139481-59-7
Molecular Formula C24H20N6O3

Specifications

Quantity 10 mg
Formula Weight 440.45
Purity >98%
Molecular Weight (g/mol) 440.463
Chemical Name or Material Candesartan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.