missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel Bioanalysis™ Cap Strips

Product Code. 11942412
Change view
Click to view available options
Quantity:
48/Pk.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11942412 48/Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11942412 Supplier Macherey-Nagel Bioanalysis™ Supplier No. 740478
  • ONLINE EXCLUSIVE PRICE
    41.25 €
    38.43 € / Pack of 1
    Save 2.82 €
    6% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cap Strips for sealing of Tube Strips or Round‐well Blocks.

  • Provided in 8-well strip
  • For use with tube strips

Specifications

Quantity 48/Pk.
Format 8-well Strip
For Use With (Equipment) Tube Strips
Temperature Range (Metric) 18°C to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.