missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAP2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92158-0.1ml
This item is not returnable.
View return policy
Description
CAP2 Polyclonal antibody specifically detects CAP2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| CAP2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| 2810452G09Rik, adenylyl cyclase-associated protein 2, CAP 2, CAP, adenylate cyclase-associated protein, 2 (yeast), PI8, SERPINB8 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 128-477 of human CAP2 (NP_006357.1). NMFNHLSAVSESIPALGWIAVSPKPGPYVKEMNDAATFYTNRVLKDYKHSDLRHVDWVKSYLNIWSELQAYIKEHHTTGLTWSKTGPVASTVSAFSVLSSGPGLPPPPPPLPPPGPPPLFENEGKKEESSPSRSALFAQLNQGEAITKGLRHVTDDQKTYKNPSLRAQGGQTQSPTKSHTPSPTSPKSYPSQKHAPVLELEGKKWRVEYQEDRNDLVISETELKQVAYIFKCEKSTIQIKGKVNSIIIDNCKKLGLVFDNVVGIVEVINSQDIQIQVMGRVPTISINKTEGCHIYLSEDALDCEIVSAKSSEMNILIPQDGDYREFPIPEQFKTAWDGSKLITEPAEIMA | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 10486 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction