Learn More
Capillary column 121-1023 DB-1 microbore fused silica 0.4μm film thickness 20m x 0.18mm J and W
Product Code. 13167484
Encompass_Preferred
Product Code. 13167484
Brand: Agilent Technologies 1211023