Learn More
Capillary column 122-1231 DB-XLB fused silica 0.1μm film thickness 30m x 0.25mm J and W
Product Code. 13184704
Encompass_Preferred
Product Code. 13184704
Brand: Agilent Technologies 1221231