Learn More
Capillary column 123-5032 DB-5 fused silica 0.25μm film thickness 30m x 0.32mm J and W
Product Code. 13197314
Encompass_Preferred
Product Code. 13197314 Supplier Agilent Technologies Supplier No. 1235032